Learn More
Thermo Scientific™ CEDIA™ Cyclosporine (CsA) Immunoassays and Calibrators
Achieve optimal low-end sensitivity and a broad dynamic range with the Thermo Scientific™ CEDIA™ Cyclosporine Plus Immunoassay.
Supplier: Thermo Scientific™ 10016283
Description
Achieve precise and accurate quantitative results crucial for monitoring drug levels of cyclosporine in kidney, liver and heart transplant patients with our easy-to-use Thermo Scientific™ CEDIA™ Cyclosporine Plus Immunoassay. Used with the Thermo Scientific CEDIA Plus High Range Calibrators, this immunoassay achieves a broad dynamic range of 25-2000ng/mL for both trough and peak samples. Applications are available on our easy-to-use Indiko™ and Indiko Plus clinical and specialty chemistry systems as well as a wide variety of other laboratory instrumentation.
Accurate:- Excellent low-end sensitivity
- Correlation with gold standard methods
- Precise throughput standard curve assay range
- Lot-to-lot consistency
Specifications
2°C to 8°C | |
Cyclosporine (CsA) | |
Reagent |
CEDIA Cyclosporine (CsA) Assay for Indiko | |
Plasma, Serum, Plasma, Serum | |
41 mL, 19 mL |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.