Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ DreamTaq Green PCR Master Mix (2X) PROMO
Save time in PCR applications with Master Mix and get higher sensitivity, longer PCR products and higher yields compared to conventional Taq DNA polymerase.
$98.00 - $436.00
Specifications
GC-Rich PCR Performance | Low |
---|---|
Polymerase | DreamTaq DNA Polymerase |
Reaction Speed | Standard |
Product Line | DreamTaq™ |
Fidelity (vs. Taq) | 1X |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
FERK1081PM
|
Thermo Scientific™
K1081 |
200 Reactions |
Each for $98.00
|
|
|||||
FERK1082PM
|
Thermo Scientific™
K1082 |
1000 Reactions |
Each for $436.00
|
|
|||||
Description
Highlights
- Direct loading of PCR product on gel
- High yields and high sensitivity of PCR
- Amplification of long targets up to 6 kb from genomic DNA and up to 20 kb from viral DNA
- Robust amplification with minimal optimization
- Incorporates modified nucleotides, but does not incorporate dUTP
Applications
- Routine PCR amplification of DNA fragments up to 6 kb
- Genotyping
- High throughput PCR
Specifications
Low | |
Standard | |
1X | |
3'-A |
DreamTaq DNA Polymerase | |
DreamTaq™ | |
No | |
SuperMix or Master Mix |
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?
Product Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title