Learn More
Research Products International Corp Pipet Box Organizer, Vertical, 4 Box Capacity of 16 1/2 Inch Long Pipet Boxes
Supplier: Research Products International Corp 145620
Clear acrylic organizers accommodate square and round stainless steel pipet boxes. Alllows box cover to be removed with one hand operation. To prevent possible warping of organizers, allow pipet cans to cool to room temperature before placing in racks. Capacity four boxes.