Learn More
Pipette Supplies, Inc. PIPETMAN SEAL P100 100UL, 5PK
SDP
Supplier:  Pipette Supplies, Inc. 90100/5
Encompass
Piston seal for:Gilson Pipetman Classic 100μL (P100)Gilson Pipetman Fixed F25, F50, F100Gilson Pipetman Neo 100μL (P100N)Alternative to Gilson F144604, F144496.With 30100, alternative to Gilson set F144864.Made in the USA
Catalog No. 50-131-7622
Explore available promotions