Learn More
Chemglass Life Sciences Pipette Canister, 15.2cm, Stainless Steel, Square
Supplier: Chemglass Life Sciences CLS1710001
Pipette Canister, 15.2cm, Stainless Steel, Square. These Pipette Canisters are made from 304 Stainless Steel and offer a high resistance to corrosion. The square cans are available in three lengths, each one 5.1cm (2 inches) square. The round canisters are 6.4cm (2-1/2”) in diameter. All Canisters come with Silicone pads on the inside of top and bottom to protect the pipettes.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.